missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ZNF291 Recombinant Protein

Brand:  Abnova™ H00049855-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189362

  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH15212
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
57.75
8
Glutathione Sepharose 4 Fast Flow
10 ug
MGLIDKLCACFLSVQGPVDENPKMAIFLQHAAGLLHAMCTLCFAVTGRSYSIFDNNRQDPTGLTAALQATDLAGVLHMLYCVLFHGTILDPSTASPKENYTQNTIQVAIQSLRFFNSFAALHLPAFQSIVGAEGLSLAFRHMASSLLGHCSQVSCESLLHEVIVCVGYFTVNHPDNQVIVQSGRHPTVLQKLCQLPFQYFSDPRLIKVLFPSLIAACYNNHQNKIILEQEMSCVLLATFIQDLAQTPGQAENQPYQPKGKCLGSQDYLELANRFPQQAWEEARQFFLKKEKK
FLJ31533/FLJ31953/KIAA1454/MSTP063/ZNF291/Zfp291
SCAPER
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
49855
ZNF291 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SCAPER
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers