missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ZNF291 Recombinant Protein
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH15212 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 49855 |
Molecular Weight (g/mol) | 57.75 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16189362
|
Abnova™
H00049855-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16199362
|
Abnova™
H00049855-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH15212 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
57.75 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SCAPER | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
49855 | |
ZNF291 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGLIDKLCACFLSVQGPVDENPKMAIFLQHAAGLLHAMCTLCFAVTGRSYSIFDNNRQDPTGLTAALQATDLAGVLHMLYCVLFHGTILDPSTASPKENYTQNTIQVAIQSLRFFNSFAALHLPAFQSIVGAEGLSLAFRHMASSLLGHCSQVSCESLLHEVIVCVGYFTVNHPDNQVIVQSGRHPTVLQKLCQLPFQYFSDPRLIKVLFPSLIAACYNNHQNKIILEQEMSCVLLATFIQDLAQTPGQAENQPYQPKGKCLGSQDYLELANRFPQQAWEEARQFFLKKEKK | |
FLJ31533/FLJ31953/KIAA1454/MSTP063/ZNF291/Zfp291 | |
SCAPER | |
Wheat Germ (in vitro) | |
GST |