missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ZCCHC6 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| Target | ZCCHC6 |
| Synonym | DKFZp666B142, DKFZp686C11112, DKFZp686F119, DKFZp686I1269, EC 2.7.7.52, FLJ13409, HS2, KIAA1711, PAP associated domain containing 6, PAPD6, terminal uridylyltransferase 7, TUT7, TUTase 7, Zinc finger CCHC domain-containing protein 6, zinc finger, CCHC domain containing 6 |
| Gene Symbol | ZCCHC6 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction