missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YOD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | YOD1 |
|---|---|
| Concentration | 0.48 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16843955
|
Proteintech
25370-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16833955
|
Proteintech
25370-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Deubiquitinating enzymes (DUBs; see MIM 603478) areproteases that specifically cleave ubiquitin (MIM 191339) linkages,negating the action of ubiquitin ligases. DUBA8 belongs to a DUBsubfamily characterized by an ovarian tumor (OTU) domain.Specifications
| YOD1 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q5VVQ6 | |
| 55432 | |
| YOD1 Fusion Protein Ag21881 | |
| Primary | |
| -20°C | |
| YOD1 |
| 0.48 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DUBA 8, DUBA8, HIN 7, HIN7, HIV 1 induced protease 7, HsHIN7, OTUD2, PRO0907, Ubiquitin thioesterase OTU1, YOD1 | |
| YOD1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title