Learn More
YOD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25370-1-AP-20UL
This item is not returnable.
View return policy
Description
Deubiquitinating enzymes (DUBs; see MIM 603478) areproteases that specifically cleave ubiquitin (MIM 191339) linkages,negating the action of ubiquitin ligases. DUBA8 belongs to a DUBsubfamily characterized by an ovarian tumor (OTU) domain.Specifications
| YOD1 | |
| Polyclonal | |
| Unconjugated | |
| YOD1 | |
| DUBA 8, DUBA8, HIN 7, HIN7, HIV 1 induced protease 7, HsHIN7, OTUD2, PRO0907, Ubiquitin thioesterase OTU1, YOD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55432 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.48 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5VVQ6 | |
| YOD1 | |
| YOD1 Fusion Protein Ag21881 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |