Learn More
YOD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25370-1-AP-20UL
 View more versions of this product
Product Code. 16843955
  • 1710.33 NOK / 20µL
Description
Deubiquitinating enzymes (DUBs; see MIM 603478) areproteases that specifically cleave ubiquitin (MIM 191339) linkages,negating the action of ubiquitin ligases. DUBA8 belongs to a DUBsubfamily characterized by an ovarian tumor (OTU) domain.
Specifications
Specifications
YOD1
Polyclonal
Unconjugated
YOD1
DUBA 8, DUBA8, HIN 7, HIN7, HIV 1 induced protease 7, HsHIN7, OTUD2, PRO0907, Ubiquitin thioesterase OTU1, YOD1
Rabbit
Antigen Affinity Chromatography
RUO
55432
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.48 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5VVQ6
YOD1
YOD1 Fusion Protein Ag21881