missing translation for 'onlineSavingsMsg'
Learn More

YIPF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15473-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16884664

  • 3468.15 NOK / 150µL
Estimated Shipment: 16-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

YIPF4
Polyclonal
Unconjugated
YIPF4
FinGER4, Nbla11189, Protein YIPF4, Yip1 domain family, member 4, YIP1 family member 4, YIPF4
Rabbit
Antigen Affinity Chromatography
RUO
362699, 67864, 84272
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5M7T4, Q8C407, Q9BSR8
Yipf4
YIPF4 Fusion Protein Ag7777
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers