missing translation for 'onlineSavingsMsg'
Learn More

YIPF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen YIPF4
Concentration 0.13 mg/mL
Applications Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16894664
View Documents
Proteintech
15473-1-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16884664
View Documents
Proteintech
15473-1-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

YIPF4
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Mouse, Human
Q5M7T4, Q8C407, Q9BSR8
362699, 67864, 84272
YIPF4 Fusion Protein Ag7777
Primary
-20°C
YIPF4
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
FinGER4, Nbla11189, Protein YIPF4, Yip1 domain family, member 4, YIP1 family member 4, YIPF4
Yipf4
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.