Learn More
XRCC5/Ku80 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16389-1-AP-150UL
This item is not returnable.
View return policy
Description
Required for activation of ATM following ionizing radiation. May act by regulating dephosphorylation of ATM.Specifications
| XRCC5/Ku80 | |
| Polyclonal | |
| Unconjugated | |
| XRCC5 | |
| 86 kDa subunit of Ku antigen, CTC85, CTCBF, DNA repair protein XRCC5, G22P2, KARP 1, KARP1, Ku80, Ku86, KUB2, NFIV, Nuclear factor IV, Thyroid lupus autoantigen, TLAA, XRCC5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7520 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13010 | |
| XRCC5 | |
| XRCC5/Ku80 Fusion Protein Ag9454 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |