Learn More
XRCC5/Ku80 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. p-7208031 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16806004 20 μL 20µL
16895994 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16806004 Supplier Proteintech Supplier No. 163891AP20UL
Rabbit Polyclonal Antibody
Required for activation of ATM following ionizing radiation. May act by regulating dephosphorylation of ATM.
Specifications
Antigen XRCC5/Ku80
Applications Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene XRCC5
Gene Accession No. P13010
Gene Alias 86 kDa subunit of Ku antigen, CTC85, CTCBF, DNA repair protein XRCC5, G22P2, KARP 1, KARP1, Ku80, Ku86, KUB2, NFIV, Nuclear factor IV, Thyroid lupus autoantigen, TLAA, XRCC5
Gene Symbols XRCC5
Host Species Rabbit
Immunogen XRCC5/Ku80 Fusion Protein Ag9454
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7520
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG