Learn More
Endecotts™ Woven Wire Mesh Sieve
Product Code. 11537494
Change view
Click to view available options
Diameter (Metric) Outer:
200 mm
11.8 in.
300 mm
450 mm
Pore Size:
40 μm
75 μm
1 mm
125 μm
150 μm
2 mm
250 μm
300 μm
425 μm
500 μm
600 μm
Unit Size:
Each
Product Code. Diameter (Metric) Outer Pore Size unitSize
11537494 200 mm 300 μm Each
11567514 200 mm 40 μm Each
11571955 200 mm 75 μm Each
15631694 200 mm 125 μm Each
11537504 200 mm 125 μm Each
11517504 200 mm 150 μm Each
11557494 200 mm 250 μm Each
11597484 200 mm 425 μm Each
15671694 200 mm 500 μm Each
11577484 200 mm 500 μm Each
11557484 200 mm 600 μm Each
11547474 200 mm 2 mm Each
12857980 300 mm 250 μm Each
12875691 450 mm 1 mm Each
15640470 11.8 in. 250 μm Each
15 options
Product Code. 11537494
Brand: Endecotts™ 200SIW.300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Available in a variety of diameters and apertures
Endecotts Woven Wire Mesh Sieves are made of stainless steel and available in a wide variety of diameters and apertures.
  • Woven Wire Mesh Sieve
  • Made of Stainless Steel
Specifications
Pore Size 300 μm
Type Sieve
Diameter (English) Outer 7.87 in.
Material Stainless Steel
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Mesh Material Woven Mesh
Sieve Number 3