Learn More
MedChemExpress Withaferin A
Specifications
Specifications
| Content And Storage | 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| Molecular Weight (g/mol) | 470.6 g/mol |
| Shipping Condition | Room temperature |
| For Use With (Application) | Cancer-Kinase/protease |
| CAS | 5119-48-2 |
| Product Type | Natural Products |
| Purity | 98.91% |
| Solubility | DMSO : 100 mg/mL (212.49 mM∣Need ultrasonic) |
| Target | NF-κB∣Ferroptosis |
| Quantity | 1 mg |