Learn More
- Home
- Products
- Stoppers and Closures
- Bottle Caps
- Fisherbrand™ White Polypropylene Caps
Fisherbrand™ White Polypropylene Caps
Product Code. 16124617 Shop All Fisherbrand Products
Change view
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
Liner Material:
PTFE/Polyethylene Foam
Polyethylene Foam
Pulp/Vinyl
Quantity:
100/Cs.
1300/Cs.
144/Cs.
2100/Cs.
288/Cs.
370/Cs.
500/Cs.
Unit Size:
Pack of 100
Pack of 1300
Pack of 144
Pack of 2100
Pack of 288
Pack of 370
Pack of 500
| Product Code. | Neck Finish | Liner Material | Quantity | unitSize |
|---|---|---|---|---|
| 16124617 | 53-400 | Polyethylene Foam | 500/Cs. | Pack of 500 |
| 16408125 | 33-400 | PTFE/Polyethylene Foam | 2100/Cs. | Pack of 2100 |
| 16164477 | 53-400 | Pulp/Vinyl | 1300/Cs. | Pack of 1300 |
| 16134597 | 18-400 | PTFE/Polyethylene Foam | 100/Cs. | Pack of 500 |
| 16124597 | 15-425 | PTFE/Polyethylene Foam | 500/Cs. | Pack of 500 |
| 16144597 | 20-400 | PTFE/Polyethylene Foam | 100/Cs. | Pack of 500 |
| 16164597 | 24-400 | PTFE/Polyethylene Foam | 100/Cs. | Pack of 500 |
| 16154597 | 22-400 | PTFE/Polyethylene Foam | 500/Cs. | Pack of 500 |
| 16174597 | 28-400 | PTFE/Polyethylene Foam | 100/Cs. | Pack of 500 |
| 16184597 | 33-400 | PTFE/Polyethylene Foam | 100/Cs. | Pack of 500 |
| 16194597 | 38-400 | PTFE/Polyethylene Foam | 500/Cs. | Pack of 500 |
| 16104607 | 43-400 | PTFE/Polyethylene Foam | 500/Cs. | Pack of 500 |
| 16114607 | 45-400 | PTFE/Polyethylene Foam | 500/Cs. | Pack of 500 |
| 16134607 | 53-400 | PTFE/Polyethylene Foam | 100/Cs. | Pack of 500 |
| 16124607 | 48-400 | PTFE/Polyethylene Foam | 100/Cs. | Pack of 100 |
| 16144607 | 58-400 | PTFE/Polyethylene Foam | 100/Cs. | Pack of 500 |
| 16154607 | 70-400 | PTFE/Polyethylene Foam | 144/Cs. | Pack of 144 |
| 16164607 | 83-400 | PTFE/Polyethylene Foam | 144/Cs. | Pack of 144 |
| 16174607 | 89-400 | PTFE/Polyethylene Foam | 144/Cs. | Pack of 144 |
| 16184607 | 63-400 | PTFE/Polyethylene Foam | 288/Cs. | Pack of 288 |
| 16478125 | 53-400 | Polyethylene Foam | 1300/Cs. | Pack of 1300 |
| 16488125 | 89-400 | Polyethylene Foam | 370/Cs. | Pack of 370 |
| 16114647 | 53-400 | Pulp/Vinyl | 288/Cs. | Pack of 288 |
Description
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| Quantity | 500/Cs. |
| Liner Material | Polyethylene Foam |
| Neck Finish | 53-400 |
| Product Type | Lined Cap |