Learn More
DWK Life Sciences Wheaton™ Preassembled Polypropylene Caps
Conveniently preassembled with septa
Brand: DWK Life Sciences 242762
Additional Details : Weight : 0.92990kg
Description
- Screw Caps
- Polypropylene
- Black
Specifications
TFE/Silicone | |
Polypropylene | |
10-425 |
Black | |
Screw Cap | |
12 mm x 32 mm E-Z Vials |