Learn More
DWK Life Sciences Wheaton™ Preassembled Polypropylene Caps
Conveniently preassembled with septa
590.00 NOK - 1545.00 NOK
Product Code | Brand | For Use With (Equipment) | Closure Size | Material (Liner) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Closure Size | Material (Liner) | Price | Quantity & Availability | |||
12458862
|
DWK Life Sciences
242760 |
12 mm x 32 mm E-Z Vials | 10-425 | TFE/Styrene-Butadiene Rubber |
590.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||
12468862
|
DWK Life Sciences
242761 |
12 mm x 32 mm E-Z Vials | 10-425 | PTFE/Silicone/PTFE |
920.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||
12478722
|
DWK Life Sciences
240602 |
12 mm x 32 mm Vials | 8-425 | PTFE/Silicone |
735.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||
11581245
|
DWK Life Sciences
242768 |
15 x 45 mm Vials | 13-425 | PTFE/Silicone |
1545.00 NOK
Pack of 100 |
Estimated Shipment: 17-04-2024
Log in to see stock. |
|||
12478862
|
DWK Life Sciences
242762 |
12 mm x 32 mm E-Z Vials | 10-425 | TFE/Silicone |
800.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||
Description
- Screw Caps
- Polypropylene
- Black
Specifications
Black | |
Polypropylene | |
Screw Cap |