Learn More
DWK Life Sciences Wheaton™ Preassembled Polypropylene Caps
Conveniently preassembled with septa
Brand:  DWK Life Sciences 242768
1467.75 NOK valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.47630kg
 View more versions of this product
Product Code. 11581245
  • 1545.00 NOK / Pack of 100
Estimated Shipment: 03-06-2024
Description
  • Screw Caps
  • Polypropylene
  • Black
Specifications
Specifications
PTFE/Silicone
Polypropylene
13-425
Black
Screw Cap
15 x 45 mm Vials