Learn More
Cytiva™ Whatman™ Qualitative Filter Paper Grade CF 12
Product Code. 11915376
Change view
Click to view available options
Quantity:
100 Pk.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11915376 100 Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11915376 Supplier Cytiva™ Supplier No. 10535097
Add to basket
Add to basket
Whatman™ Grade CF 12 qualitative filter paper is ideal for general laboratory applications, such as removing particulate matter, clarifying liquids and separating solids from liquids in qualitative analysis. The Grade CF 12 offers medium retention and flow rate.
  • Qualitative filter paper sheets
  • Grade CF 12
  • Ideal for general filtration applications
  • Medium retention and flow rate
  • Suitable for various industries including pharmaceutical, environmental and food testing
TRUSTED_SUSTAINABILITY
Specifications
Description Qualitative filter paper
Product Line Whatman
Product Type Qualitative Filter Paper
Quantity 100 Pk.
Material Cellulose fibers
Format Sheets 210 x 297 mm
Purity or Quality Grade Grade CF 12
For Use With (Application) Suitable for various industries including pharmaceutical, environmental and food testing