Learn More
IBA Lifesciences WET FRED Set
Shop All IBA Lifesciences Products
Click to view available options
Capacity (Metric):
1 mL Gravity Flow Columns
5 to 10 mL Gravity Flow Columns
Unit Size:
1 set
Description
- WET FRED enables the application of large cell culture supernatant or lysate volumes to a gravity flow column in a simple way
- Based on the siphon principle, it connects the gravity flow column with the bottle containing culture supernatant, lysate, or buffer that are to be applied
Specifications
Specifications
| Capacity (Metric) | 1 mL Gravity Flow Columns |
| Quantity | 1 Set |
| For Use With (Application) | Eases Protein Purification |
| Type | Protein Purification Aid |
For Research Use Only.