missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR76 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25528-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
WDR76 | |
Polyclonal | |
Unconjugated | |
WDR76 | |
CDW14, WD repeat domain 76, WDR76 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79968 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H967 | |
WDR76 | |
WDR76 Fusion Protein Ag22114 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |