missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR32 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20483-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Proteintech's WDR32 antibody 20483-1-AP primarily detects WDR32 isoform 1 with a theoretical MW of 61 kDa.
DCAF10 may function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex.Specifications
WDR32 | |
Polyclonal | |
Unconjugated | |
DCAF10 | |
DCAF10, WD repeat domain 32, WDR32 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
242418, 313242, 79269 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2AKB9, Q5QP82 | |
Dcaf10 | |
WDR32 Fusion Protein Ag14360 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |