missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR32 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | WDR32 |
---|---|
Concentration | 0.18 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890025
|
Proteintech
20483-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16880025
|
Proteintech
20483-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Proteintech's WDR32 antibody 20483-1-AP primarily detects WDR32 isoform 1 with a theoretical MW of 61 kDa.
DCAF10 may function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex.Specifications
WDR32 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
A2AKB9, Q5QP82 | |
242418, 313242, 79269 | |
WDR32 Fusion Protein Ag14360 | |
Primary | |
-20°C | |
DCAF10 |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DCAF10, WD repeat domain 32, WDR32 | |
Dcaf10 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |