Learn More
BINDER™ Water Supply
Water Supply for refillable tubular bags including peristaltic tubing, connectors, and fine gas filters.
Brand:  BINDER™ 8009-1058
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 16454019
  • 3255.00 NOK / Each
Estimated Shipment: 15-07-2024
Description
  • Water supply for versions of CO2 Incubators with active humidification
  • Sets consist of empty, refillable tubular bags including peristaltic tubing, connectors, and fine gas filters
Specifications
Specifications
Water supply - Set consisting of 12 empty, refillable tubular bags including peristaltic tubing, connectors, and fine gas filters
Plastic
Set consisting of 12 empty, refillable tubular bags including peristaltic tubing, connectors, and fine gas filters
BINDER Chambers CBF Series CO2 Incubators with Hot Air Sterilization and Humidity Regulation
Water Supply