Learn More
BINDER™ Water Supply
Water Supply for refillable tubular bags including peristaltic tubing, connectors, and fine gas filters.
Brand: BINDER™ 8009-1058
Additional Details : Weight : 0.00100kg
Description
- Water supply for versions of CO2 Incubators with active humidification
- Sets consist of empty, refillable tubular bags including peristaltic tubing, connectors, and fine gas filters
Specifications
Water supply - Set consisting of 12 empty, refillable tubular bags including peristaltic tubing, connectors, and fine gas filters | |
Plastic | |
Set consisting of 12 empty, refillable tubular bags including peristaltic tubing, connectors, and fine gas filters |
BINDER Chambers CBF Series CO2 Incubators with Hot Air Sterilization and Humidity Regulation | |
Water Supply |