Learn More
BINDER™ Water Supply
Water Supply for refillable tubular bags including peristaltic tubing, connectors, and fine gas filters.
930.00 NOK - 3255.00 NOK
Specifications
For Use With (Equipment) BINDER Chambers CBF Series CO2 Incubators with Hot Air Sterilization and Humidity Regulation
Material Plastic
Type Water Supply
View More Specs
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
16464019
View Documents Promotion Details
BINDER™
8009-1083
Set consisting of 3 empty, refillable tubular bags including peristaltic tubing, connectors, and fine gas filters
930.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
16454019
View Documents Promotion Details
BINDER™
8009-1058
Set consisting of 12 empty, refillable tubular bags including peristaltic tubing, connectors, and fine gas filters
3255.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
Description
  • Water supply for versions of CO2 Incubators with active humidification
  • Sets consist of empty, refillable tubular bags including peristaltic tubing, connectors, and fine gas filters
Specifications
Specifications
BINDER Chambers CBF Series CO2 Incubators with Hot Air Sterilization and Humidity Regulation
Plastic
Water Supply