Learn More
Miele Professional™ Lab Washer
Analysis grade conditioning of laboratory glassware.
POA
Specifications
Product Type | Professional Washer/Disinfector |
---|---|
For Use With (Application) | Disinfection of laboratory instruments |
Dimensions (D x W x H) Interior | 60 x 30 x 83.5 cm |
Electrical Requirements | 400V 50Hz 3N AC 9.3 kW |
Description
- Freestanding device (with lid), for undercounter use
- Integrated dispenser pumps for liquid process chemicals (neutralizing- / cleaning agent)
- Touch Control with 14 programmes
- Four connections for 2 x cold, hot and demineralised water
- Powerful circulation pump, max. throughput 400L/ min
Specifications
Professional Washer/Disinfector | |
Disinfection of laboratory instruments | |
60 x 30 x 83.5 cm | |
400V 50Hz 3N AC 9.3 kW |