Learn More
R&L Enterprises™ Ward Pattern Furnace Tongs
Product Code. 11899601 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
590 mm
740 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11899601 590 mm Each
11869701 740 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11899601
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
For protection and safety when handling heated objects in the laboratory. R&L Enterprises™ Ward Pattern Furnace Tongs are fitted with a circular shield to protect the hand against radiation from the furnace.
  • Manufactured from stainless steel
  • For handling a wide variety of crucibles, especially in muffle furnaces
  • Operated by thumb action with one hand
Specifications
Product Type Furnace Tongs
For Use With (Application) For handling a wide variety of crucibles and to protect the hand against radiation from the furnace
Quantity 1 Ea.
Length (Metric) 590 mm
Material Stainless Steel