Learn More
Fisherbrand™ Wall Bracket for Additional Cartridge
Artikelnummer. 11974159 Alle ansehen Fisherbrand Traceable Produkte
Ansicht ändern
Click to view available options
Quantity:
1 Each
Packungsgröße:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
11974159 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 11974159
  • ONLINE EXCLUSIVE PRICE
    1071.00 NOK
    703.59 NOK / Pack of 1
    Save 367.41 NOK
    34% Off
Alle Werbeaktionen anzeigen
 Restricted Item
Estimated Shipment: 15-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
For adding additional light water filtration and purification cartridges. Fisherbrand™ Wall Bracket for Additional Cartridge provides for safe and secure connection.
  • Facilitates the water purification process
  • Supplied in individual packs
Spezifikation
Filter Size 60 x 11 cm
For Use With (Application) Water purification
For Use With (Equipment) Deionization Cartridges; Activated Carbon Cartridge; Micron Filter Cartridge; TOC Ion Exchange Cartridge; KCR Quick Coupling Kit
Quantity 1 Each
Type Wall Bracket