Learn More
Fisherbrand™ Wall Bracket for Single Cartridge
Product Code. 11964159 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11964159 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11964159
  • ONLINE EXCLUSIVE PRICE
    1336.20 NOK
    879.35 NOK / Pack of 1
    Save 456.85 NOK
    34% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
For affixing a single cartridge or the first in a series of cartridges. Fisherbrand™ Wall Brackets for Single Cartridges provide for safe and secure connection.
  • Facilitates the water purification and filtration process
  • Aluminum and plastic construction with tap input
  • Delivered with 2m of tube, screws and dowels
Specifications
Filter Size 65 x 11 cm
For Use With (Application) For water purification
For Use With (Equipment) For Deionization Cartridges; Activated Carbon Cartridge; Micron Filter Cartridge; TOC Ion Exchange Cartridge; KCR Quick Coupling Kit
Includes Valve on the top of the support
Quantity 1 Each
Type Wall Bracket