missing translation for 'onlineSavingsMsg'
Learn More

VPS11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 19140-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16848894

  • 3828.53 NOK / 150µL
Estimated Shipment: 17-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps11 protein. The mammalian class C Vps proteins are predomitly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

VPS11
Polyclonal
Unconjugated
VPS11
END1, hVPS11, PEP5, RING finger protein 108, RNF108, VPS11
Rabbit
Antigen Affinity Chromatography
RUO
315600, 55823, 71732
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q91W86, Q9H270
Vps11
VPS11 Fusion Protein Ag6227
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers