missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | VPS11 |
|---|---|
| Concentration | 0.17 mg/mL |
| Applications | Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16858894
|
Proteintech
19140-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16848894
|
Proteintech
19140-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps11 protein. The mammalian class C Vps proteins are predomitly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. Alternative splicing results in multiple transcript variants.Specifications
| VPS11 | |
| Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q91W86, Q9H270 | |
| 315600, 55823, 71732 | |
| VPS11 Fusion Protein Ag6227 | |
| Primary | |
| -20°C | |
| VPS11 |
| 0.17 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| END1, hVPS11, PEP5, RING finger protein 108, RNF108, VPS11 | |
| Vps11 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title