missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps11 protein. The mammalian class C Vps proteins are predomitly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | VPS11 |
| Applications | Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry, Flow Cytometry |
| Classification | Polyclonal |
| Concentration | 0.17 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | VPS11 |
| Gene Accession No. | Q91W86, Q9H270 |
| Gene Alias | END1, hVPS11, PEP5, RING finger protein 108, RNF108, VPS11 |
| Gene Symbols | Vps11 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?