Learn More
Fisherbrand™ Vortex Mixers Tube Holders
Product Code. 15746959 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Positions:
1
18
24
Unit Size:
Each
Product Code. No. of Positions unitSize
15746959 24 Each
15776959 1 Each
15756959 18 Each
3 options
Product Code. 15746959
Only null left
Add to basket
Add to basket
For use with Fisherbrand vortex mixers
  • Mount atop mixer for single-tube, hands-free mixing
  • Easily attach without tools
TRUSTED_SUSTAINABILITY
Specifications
Holds 0.5 mL
Product Type Microtube Holder
Dimensions (L x W) 9 x 9 in. (229 x 229 mm)
Length (English) 9 in.
Length (Metric) 229 mm
Width (English) 9 in.
Width (Metric) 229 mm
For Use With (Equipment) Vortex mixer
No. of Positions 24