missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VN1R3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24659-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Putative pheromone receptor. There seems to be a deletion in the gene coding for this protein in about 98% of the population. The gorilla and orangutan orthologous proteins do not exist, their genes are pseudogenes.Specifications
VN1R3 | |
Polyclonal | |
Unconjugated | |
VN1R3 | |
FKSG46, V1RL3, V1RL3p, VN1R3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
317702 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BXE9 | |
VN1R3 | |
VN1R3 Fusion Protein Ag20382 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |