missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VN1R3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | VN1R3 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803155
|
Proteintech
24659-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16893145
|
Proteintech
24659-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Putative pheromone receptor. There seems to be a deletion in the gene coding for this protein in about 98% of the population. The gorilla and orangutan orthologous proteins do not exist, their genes are pseudogenes.Specifications
VN1R3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q9BXE9 | |
317702 | |
VN1R3 Fusion Protein Ag20382 | |
Primary | |
-20°C | |
VN1R3 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FKSG46, V1RL3, V1RL3p, VN1R3 | |
VN1R3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |