Learn More
MP Biomedicals Vitamin D Control Diet, Powder
Product Code. 11493011 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 kg
20 kg
50 kg
Unit Size:
10kg
20kg
50kg
Product Code. Quantity unitSize
11493011 10 kg 10kg
11403021 20 kg 20kg
11413021 50 kg 50kg
3 options
Product Code. 11493011
Brand: MP Biomedicals 0296022510
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Vitamin D Control Diet - 5 IU/gm.
Vitamin D control diet contains all nutrients required for rats and provides growth and reproduction equal to that of commercial stock diets. Using whole wheat as the source of carbohydrate the rats experience better growth and development than with mono or disaccharides. The calcium content of the diet can be varied by changing the amount of CaCO3 added. The phosphorous content can be increased from approximately 0.35% (the wheat flour contains most of the phosphorous and the casein a small and variable amount) by the addition of potassium diphosphate.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Product Type Animal Diet
Disposable Single-use
Format Powder
Quantity 10 kg
Sterility Non-sterile
Storage Requirements Room Temperature
Length (English) 1 ft.
Length (Metric) 0.3048 m
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.