Learn More
Fisherbrand™ C-Shape Vinyl Coated Lead Rings
Product Code. 11736725 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
Flasks (1 to 4 L)
Flasks (125 to 500 mL)
Flasks (250 to 1000 mL)
Flasks (500 to 2000 mL)
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
11736725 Flasks (250 to 1000 mL) Each
11726725 Flasks (125 to 500 mL) Each
11756725 Flasks (1 to 4 L) Each
11746725 Flasks (500 to 2000 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11736725
  • ONLINE EXCLUSIVE PRICE
    426.00 NOK
    376.97 NOK / Pack of 1
    Save 49.03 NOK
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
Stabilize vessels and labware or immerse in water baths. Fisherbrand™ Vinyl-coated Lead Rings keep flasks and other labware weighted down without causing damage.
  • Vinyl-coated lead rings will not mark or scratch benchtops and labware
  • C-shape format allows some manual size variation for a customized fit
Specifications
Autoclavable Not Autoclavable
Color Blue
Holds Flasks (250 to 1000 mL)
Material Lead
Positions 1
Shape Circular
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) Flasks
Quantity 1 Ea.
Size 250 to 1000 mL
Diameter (Metric) Outer 90 mm
Weight (Metric) 0.472 kg
Product Type Lead Ring
Diameter (Metric) Inner 51 mm
Shelves None