Learn More
Gilson™ VERITY 4220 Dual Syringe Pump
Product Code. 16616165 Shop All Gilson Products
Change view
Click to view available options
Temperature (Metric) Max:
40°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Max unitSize
16616165 40°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16616165 Supplier Gilson™ Supplier No. 31130003
  • ONLINE EXCLUSIVE PRICE
    115820.00 NOK
    102861.71 NOK / Pack of 1
    Save 12958.29 NOK
    11% Off
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
The VERITY™ Syringe Pumps provide speed and reliability to small- and large-volume, repetitive liquid transfers in HPLC, SPE, and other general liquid handling methods. These pumps work with many VERITY systems and are available in single syringe, dual syringe, and dual syringe with tee configurations. A pressure sensor is available in some models.
  • Choose the Right Pump for Your Needs
  • VERITY 4000-Series Syringe Pumps are ideal for all VERITY GX-series liquid handlers and the 223 Sample Changer.
  • VERITY 4060 and 4260 Syringe Pumps are designed for use with VERITY ASPEC™ SPE systems to automate liquid handling and extractions using positive pressure elution.
  • The VERITY 4060 Single Syringe Pump and VERITY 4260 Dual Syringe Pump when paired with an ASPEC system, can automate liquid handling procedures and solid phase extraction (SPE) using positive pressure elution.
Specifications
Voltage 100/240 V
Type Dual Syringe Pump
Weight (Metric) 7.1 kg
Dimensions (L x W x H) Exterior 22.6 x 17.1 x 26.9 cm
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 5°C
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.