Learn More
Eppendorf™ Varispenser™ 2x Bottle-Top Dispenser with Recirculation Valve
Ideal for dispensing aliquots of liquid from large supply bottles.
4075.00 NOK - 7395.00 NOK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15787367
|
Eppendorf™
4967000014 |
0.2 to 2.0 mL |
4075.00 NOK
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
15797367
|
Eppendorf™
4967000022 |
0.5 to 5.0 mL |
4075.00 NOK
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
15707377
|
Eppendorf™
4967000030 |
1.0 to 10 mL |
4075.00 NOK
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
15717377
|
Eppendorf™
4967000049 |
2.5 to 25 mL |
5450.00 NOK
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
15781587
|
Eppendorf™
4967000057 |
5.0 to 50 mL |
5895.00 NOK
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
15727377
|
Eppendorf™
4967000065 |
10 to 100 mL |
7395.00 NOK
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Description
Ideal for dispensing aliquots of liquid from large supply bottles.
- Recirculation valve
- Standard GL 45 thread on all sizes for most common bottle threads
- Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
- Screwable air vent cap for easy assembly of drying tube or microfilter
- Oval-shaped dispenser housing for easy transportation
- PFA seal of the sliding piston prevents seizing up
- Fixation slide on interior toothed track for rapid and accurate volume adjustment
- High chemical resistance of all dispensing components
- Telescopic aspirating tubes for variable adjustment to bottle height
- Fully autoclavable without disassembly
- User adjustment option
Specifications
1 |