Learn More
Eppendorf™ Varispenser™ 2x Bottle-Top Dispenser with Recirculation Valve
Shop All Eppendorf Products
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
0.2 to 2.0 mL
0.5 to 5.0 mL
1.0 to 10 mL
10 to 100 mL
2.5 to 25 mL
5.0 to 50 mL
missing translation for 'unitSize'
Each
Description
Ideal for dispensing aliquots of liquid from large supply bottles.
- Recirculation valve
- Standard GL 45 thread on all sizes for most common bottle threads
- Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
- Screwable air vent cap for easy assembly of drying tube or microfilter
- Oval-shaped dispenser housing for easy transportation
- PFA seal of the sliding piston prevents seizing up
- Fixation slide on interior toothed track for rapid and accurate volume adjustment
- High chemical resistance of all dispensing components
- Telescopic aspirating tubes for variable adjustment to bottle height
- Fully autoclavable without disassembly
- User adjustment option
Specifications
Specifications
| Product Type | Bottletop Dispenser with Adapters |
| Channels | 1 |
| Volume (Metric) | 0.2 to 2.0 mL |
| Includes | Recirculation valve, adapters GL 25, GL 28/S 28, GL 32, GL 38, S 40, and telescopic intake tube (125 – 240 mm) |