Learn More
Cole-Parmer™ VapLock™ Threaded Plugs
Click to view available options
Material:
PTFE
Thread Size (English):
1/4 in.-28 UNF(M)
Unit Size:
Pack of 10
Description
Seal unused manifold and bottle cap ports
Warranty
None
Specifications
Specifications
| For Use With (Equipment) | VapLock Solvent Containment Systems |
| Material | PTFE |
| Thread Size (English) | 1/4 in.-28 UNF(M) |
| For Use With (Application) | Solvent Safety |
| Type | Port Plug |