Learn More
Fisherbrand™ Vacuum Regulator with Dial Gauge
Product Code. 15549632 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Includes:
with DN8 hose nozzle for 8mm ID vacuum hose
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
15549632 with DN8 hose nozzle for 8mm ID vacuum hose Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15549632
  • ONLINE EXCLUSIVE PRICE
    2835.00 NOK
    2767.19 NOK / Pack of 1
    Save 67.81 NOK
    2% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
Designed for use with Fisherbrand Chemical Duty Diaphragm Vacuum Pumps. Fisherbrand™ Vacuum Regulators feature regulator valve for intake that allows the adjustment of the ultimate pressure.
  • With DN8 hose nozzle for 8mm I.D. vacuum hose
  • Regulator valve for intake allows the adjustment of the ultimate pressure
  • Dial gauge indicates the vacuum level
  • Air regulator valve mounted directly on the pump
  • For Chemical Duty Diaphragm Vacuum Pump Cat. No 10287097
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For Fisherbrand Chemical Duty Diaphragm Vacuum Pump, Cat. No 10287097
Includes with DN8 hose nozzle for 8mm ID vacuum hose
Product Type Model DBR-B Vacuum Regulator with Dial Gauge