Learn More
Memmert™ Vacuum Oven Thermoshelf
Product Code. 11901459 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
Model 200
Model 400
Model 500
Material:
Aluminum
Stainless Steel
Unit Size:
Each
Product Code. 11901459
Brand: Memmert™ VO49B00734
Only null left
Specifications
Certifications/Compliance Calibration certificate
For Use With (Equipment) Model 400
Type Shelf
For Use With (Application) Individual overtemperature protection for each shelf MLOP (Multi-Level-Overtemperature-Control)
Includes Local Temperature Sensing (Pt100, 4-Wire-Circuit)
Material Stainless Steel