Specifications
| Certifications/Compliance | Calibration certificate |
| For Use With (Equipment) | Model 500 |
| Type | Shelf |
| For Use With (Application) | Individual overtemperature protection for each shelf MLOP (Multi-Level-Overtemperature-Control) |
| Includes | Local Temperature Sensing (Pt100, 4-Wire-Circuit) |
| Material | Aluminum |