missing translation for 'onlineSavingsMsg'
Learn More

USP36 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14783-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16893594

  • 4593.76 NOK / 150µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

May be required for maintaining multiple types of adult stem cells. May function as a transcriptional repressor by continually deubiquiting histone H2B at the promoters of genes critical for cellular differentiation, thereby preventing histone H3 ′Lys-4′ trimethylation (H3K4). Nagase T., DNA Res. 7:143-150(2000). The MGC Project Team, Genome Res. 14:2121-2127(2004). Ota T., Nat. Genet. 36:40-45(2004).
Specifications

Specifications

USP36
Polyclonal
Unconjugated
USP36
Deubiquitinating enzyme 36, DUB1, KIAA1453, Ubiquitin thiolesterase 36, USP36
Rabbit
Antigen Affinity Chromatography
RUO
57602, 72344
-20°C
Liquid
Immunoprecipitation, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B1AQJ2, Q9P275
USP36
USP36 Fusion Protein Ag6411
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers