missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP36 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | USP36 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803604
|
Proteintech
14783-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16893594
|
Proteintech
14783-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
May be required for maintaining multiple types of adult stem cells. May function as a transcriptional repressor by continually deubiquiting histone H2B at the promoters of genes critical for cellular differentiation, thereby preventing histone H3 ′Lys-4′ trimethylation (H3K4). Nagase T., DNA Res. 7:143-150(2000). The MGC Project Team, Genome Res. 14:2121-2127(2004). Ota T., Nat. Genet. 36:40-45(2004).Specifications
USP36 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
B1AQJ2, Q9P275 | |
57602, 72344 | |
USP36 Fusion Protein Ag6411 | |
Primary | |
-20°C | |
USP36 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Deubiquitinating enzyme 36, DUB1, KIAA1453, Ubiquitin thiolesterase 36, USP36 | |
USP36 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |