missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USHBP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17226-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
USHBP1, also named as Mutated in colon cancer protein 2 or AIE-75-binding protein, is a 703 amino acid protein, which belongs to the MCC family. Highest level of expression is in heart, and moderate to low expression in skeletal muscle, kidney, liver, small intestine, placenta and lung.Specifications
USHBP1 | |
Polyclonal | |
Unconjugated | |
USHBP1 | |
AIE 75 binding protein, AIEBP, MCC 2, MCC2, USH1C binding protein 1, USHBP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234395, 83878 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N6Y0, Q8R370 | |
USHBP1 | |
USHBP1 Fusion Protein Ag11064 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |