missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USHBP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | USHBP1 |
|---|---|
| Concentration | 0.31 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16807084
|
Proteintech
17226-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16897074
|
Proteintech
17226-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USHBP1, also named as Mutated in colon cancer protein 2 or AIE-75-binding protein, is a 703 amino acid protein, which belongs to the MCC family. Highest level of expression is in heart, and moderate to low expression in skeletal muscle, kidney, liver, small intestine, placenta and lung.Specifications
| USHBP1 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q8N6Y0, Q8R370 | |
| 234395, 83878 | |
| USHBP1 Fusion Protein Ag11064 | |
| Primary | |
| -20°C | |
| USHBP1 |
| 0.31 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| AIE 75 binding protein, AIEBP, MCC 2, MCC2, USH1C binding protein 1, USHBP1 | |
| USHBP1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title