Learn More
Dostmann Electronic™ USB Cable
Click to view available options
Length (Metric):
1000 mm
Unit Size:
Each
Specifications
Specifications
| Length (Metric) | 1000 mm |
| Length (Metric) Cable | 1000 mm |
| Weight (Metric) | 100 g |
| Product Type | USB Cable |
| For Use With (Equipment) | P700 - Universal-Thermometer, 1-Kanal, P705 - Universal-Thermometer, 2-Kanal, P710 - Universal-Thermometer, 1-Kanal, P715 - Universal-Thermometer, 2-Kanal, P750 Temperature-humidity instrument, P755 multi-function instrument for temperature-humidity-airfl |