Learn More
Fisherbrand™ Upright ULT Freezer Racks
Shop All Fisherbrand Products
Click to view available options
Holds:
12 (3in.) boxes
12 Boxes (2 in.)
15 (2 in.) boxes
16 (2 in.) boxes
20 (2 in.) boxes
24 (2 in.) boxes
25 (2 in.) boxes
8 (100mm) boxes
9 (2 in.) boxes
Boxes (2 or 3 in.)
Dimensions (L x W x H):
415 x 140 x 168 mm
415 x 140 x 225 mm
415 x 140 x 280 mm
550 x 140 x 205 mm
550 x 140 x 225 mm
550 x 140 x 280 mm
550 x 140 x 321 mm
550 x 143 x 315 mm
679 x 140 x 225 mm
679 x 140 x 280 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Side Access Freezer Rack |
| Holds | Boxes (2 or 3 in.) |
| Material | Stainless Steel |
| For Use With (Application) | 2 in. or 3 in. cryoboxes |
| For Use With (Equipment) | Upright ULT Freezer |
| Dimensions (L x W x H) | 550 x 143 x 315 mm |