Learn More
Fisherbrand™ Upright ULT Freezer Drawer Racks
Product Code. 15789549 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
12 Boxes (2 in.)
15 (2in.) boxes
16 (2in.) boxes
20 (2in.) boxes
24 (2in.) boxes
9 (2in.) boxes
Dimensions (L x W x H):
419 x 140 x 181 mm
419 x 140 x 240 mm
419 x 140 x 300 mm
559 x 140 x 240 mm
559 x 140 x 300 mm
559 x 140 x 359 mm
679 x 140 x 240 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Dimensions (L x W x H) Unit Size
15789549 20 (2in.) boxes 679 x 140 x 240 mm Each
15729549 9 (2in.) boxes 419 x 140 x 181 mm Each
15739549 12 Boxes (2 in.) 419 x 140 x 240 mm Each
15749549 15 (2in.) boxes 419 x 140 x 300 mm Each
15759549 16 (2in.) boxes 559 x 140 x 240 mm Each
15769549 20 (2in.) boxes 559 x 140 x 300 mm Each
15779549 24 (2in.) boxes 559 x 140 x 359 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15789549
  • ONLINE EXCLUSIVE PRICE
    1505.00 NOK
    1329.25 NOK / Pack of 1
    Save 175.75 NOK
    11% Off
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
Fisherbrand inventory control racks are ideal for all cold storage application. They fit all standard types of upright ULT freezers.
  • Holds standard cryoboxes
  • Drawers offer easy-access to the boxes
  • Made of high quality stainless steel
Specifications
Holds 20 (2in.) boxes
Material Stainless Steel
For Use With (Application) Standard 2 in. Cryoboxes
For Use With (Equipment) Upright ULT Freezer
Dimensions (L x W x H) 679 x 140 x 240 mm
Product Type Drawer Rack