missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPK1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25275-1-AP-20UL
This item is not returnable.
View return policy
Description
Component of the asymmetric unit membrane (AUM); a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. May play an important role in normal bladder epithelial physiology, possibly in regulating membrane permeability of superficial umbrella cells or in stabilizing the apical membrane through AUM/cytoskeletal interactions.Specifications
| UPK1A | |
| Polyclonal | |
| Unconjugated | |
| UPK1A | |
| Tetraspanin 21, Tspan 21, TSPAN21, UP1A, UPIA, UPK1A, UPKA, uroplakin 1A, Uroplakin Ia | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109637, 11045 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00322, Q9D132 | |
| Upk1a | |
| UPK1A Fusion Protein Ag17356 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction